CH4005798740 ChemBridge Corporation

2-(2-pyridinylamino)-8-quinolinol

  • MDL number MFCD00168938
  • Cas Number : 70125-23-4
  • Molecular Weight : 237.2566

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5175087

Safety Information

MSDS https://www.hit2lead.com/sds/5175087_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart