CH4005798744 ChemBridge Corporation

2-hydrazino-5,7-dimethylquinolin-8-ol

  • MDL number MFCD00168948
  • Cas Number : 317375-38-5
  • Molecular Weight : 203.2404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5175097

Safety Information

MSDS https://www.hit2lead.com/sds/5175097_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart