CH4005798791 ChemBridge Corporation

2,5,7-trichloroquinolin-8-ol

  • MDL number MFCD00552678
  • Cas Number : 101870-58-0
  • Molecular Weight : 248.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5175177

Safety Information

MSDS https://www.hit2lead.com/sds/5175177_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart