CH4005798934 ChemBridge Corporation

2,3-dihydrobenzo[g]quinolin-4(1H)-one

  • MDL number MFCD00552977
  • Cas Number : 21516-07-4
  • Molecular Weight : 197.2325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5175572

Safety Information

MSDS https://www.hit2lead.com/sds/5175572_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart