CH4005804386 ChemBridge Corporation

1-(3-mercaptopropanoyl)-6-methylpiperidine-2-carboxylic acid

  • MDL number MFCD00564081
  • Cas Number : 174909-66-1
  • Molecular Weight : 231.3119

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5219863

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/5219863_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart