CH4005806999 ChemBridge Corporation

methyl 3-amino-4,5,6-trimethylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00440677
  • Cas Number : 319491-25-3
  • Molecular Weight : 250.3168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5231471

Safety Information

MSDS https://www.hit2lead.com/sds/5231471_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart