CH4005807766 ChemBridge Corporation

4-(1-adamantyl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD00188088
  • Cas Number : 201992-89-4
  • Molecular Weight : 248.387

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5235261

Safety Information

MSDS https://www.hit2lead.com/sds/5235261_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart