CH4005809751 ChemBridge Corporation

1-(5-bromo-2-furoyl)piperidine

  • MDL number MFCD00460880
  • Cas Number : 57785-36-1
  • Molecular Weight : 258.1118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5245801

Safety Information

MSDS https://www.hit2lead.com/sds/5245801_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart