CH4005809764 ChemBridge Corporation

N-benzyl-5-bromo-2-furamide

  • MDL number MFCD00454467
  • Cas Number : 117845-23-5
  • Molecular Weight : 280.1173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5245842

Safety Information

MSDS https://www.hit2lead.com/sds/5245842_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart