CH4005810528 ChemBridge Corporation

5-bromo-3-ethoxy-2-hydroxybenzaldehyde

  • MDL number MFCD00579044
  • Cas Number : 20035-44-3
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5249942

Safety Information

MSDS https://www.hit2lead.com/sds/5249942_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart