CH4005810566 ChemBridge Corporation

2-(3-bromophenyl)-1,3-benzoxazol-5-amine

  • MDL number MFCD00445929
  • Cas Number : 293737-81-2
  • Molecular Weight : 289.1274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5250071

Safety Information

MSDS https://www.hit2lead.com/sds/5250071_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart