CH4005811612 ChemBridge Corporation

5,7-dichloro-2,1,3-benzoxadiazol-4-amine

  • MDL number MFCD00168460
  • Cas Number : 330982-41-7
  • Molecular Weight : 204.0135

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5256245

Safety Information

MSDS https://www.hit2lead.com/sds/5256245_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart