CH4005814623 ChemBridge Corporation

N-(2-methoxybenzyl)-3-oxobutanamide

  • MDL number MFCD00844470
  • Cas Number : 331713-77-0
  • Molecular Weight : 221.2524

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5274303

Safety Information

MSDS https://www.hit2lead.com/sds/5274303_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart