CH4005817136 ChemBridge Corporation

N-mesityl-3-oxobutanamide

  • MDL number MFCD00453329
  • Cas Number : 19359-16-1
  • Molecular Weight : 219.2796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5303985

Safety Information

MSDS https://www.hit2lead.com/sds/5303985_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart