CH4005821316 ChemBridge Corporation

1-(1-hydroxycyclohexyl)-3-(4-methoxyphenyl)propan-1-one

  • MDL number MFCD00764053
  • Cas Number : 312318-69-7
  • Molecular Weight : 262.3441

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5325517

Safety Information

MSDS https://www.hit2lead.com/sds/5325517_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart