CH4005821845 ChemBridge Corporation

2-bromo-N-isopropylbenzamide

  • MDL number MFCD00779656
  • Cas Number : 64141-90-8
  • Molecular Weight : 242.1124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5328478

Safety Information

MSDS https://www.hit2lead.com/sds/5328478_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart