CH4005822505 ChemBridge Corporation

4-bromo-N-(4-phenylbutyl)benzamide

  • MDL number MFCD00813650
  • Cas Number : 415694-66-5
  • Molecular Weight : 332.2349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5334050

Safety Information

MSDS https://www.hit2lead.com/sds/5334050_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart