CH4005823968 ChemBridge Corporation

tricyclo[3.3.1.1~3,7~]dec-2-ylideneacetic acid

  • MDL number MFCD01825775
  • Cas Number : 25220-07-9
  • Molecular Weight : 192.2542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5344646

Safety Information

MSDS https://www.hit2lead.com/sds/5344646_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart