CH4005825009 ChemBridge Corporation

2-(4-hydroxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD00687527
  • Cas Number : 6952-34-7
  • Molecular Weight : 265.2634

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5349475

Safety Information

MSDS https://www.hit2lead.com/sds/5349475_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart