CH4005825438 ChemBridge Corporation

methyl 2-(4-aminophenyl)quinoline-4-carboxylate

  • MDL number MFCD00814379
  • Cas Number : 94541-55-6
  • Molecular Weight : 278.3053

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5351631

Safety Information

MSDS https://www.hit2lead.com/sds/5351631_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart