CH4005830831 ChemBridge Corporation

(5Z)-5-(3-ethoxy-4-hydroxybenzylidene)-1,3-thiazolidine-2,4-dione

  • MDL number MFCD00683374
  • Cas Number : 1055980-74-9
  • Molecular Weight : 265.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5404527

Safety Information

MSDS https://www.hit2lead.com/sds/5404527_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart