CH4005835786 ChemBridge Corporation

1-(4-fluorophenyl)-3-(2-hydroxyphenyl)propane-1,3-dione

  • MDL number MFCD20502979
  • Cas Number : 135276-47-0
  • Molecular Weight : 258.2444

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5474563

Safety Information

MSDS https://www.hit2lead.com/sds/5474563_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart