CH4005838384 ChemBridge Corporation

5-[(4-methylphenoxy)methyl]-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01043766
  • Cas Number : 84138-77-2
  • Molecular Weight : 221.2788

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5491809

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5491809
Availability details for
Quantity
  • Availability
  • Add to Cart