CH4005838687 ChemBridge Corporation

4-chloro-N-(3-hydroxyphenyl)-3-nitrobenzamide

  • MDL number MFCD01004921
  • Cas Number : 416887-71-3
  • Molecular Weight : 292.6746

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5509882

Safety Information

MSDS https://www.hit2lead.com/sds/5509882_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart