CH4005839156 ChemBridge Corporation

4,8-dimethoxy-1-naphthaldehyde

  • MDL number MFCD00967522
  • Cas Number : 69833-11-0
  • Molecular Weight : 216.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5522241

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5522241
Availability details for
Quantity
  • Availability
  • Add to Cart