CH4005839223 ChemBridge Corporation

2-[(5-methyl-1H-benzimidazol-2-yl)amino]ethanol

  • MDL number MFCD00967560
  • Cas Number : 121477-75-6
  • Molecular Weight : 191.2297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5522535

Safety Information

MSDS https://www.hit2lead.com/sds/5522535_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart