CH4005840787 ChemBridge Corporation

2-(3,4-dimethoxyphenyl)-N-(pyridin-4-ylmethyl)ethanamine

  • MDL number MFCD01135673
  • Cas Number : 331970-88-8
  • Molecular Weight : 272.3422

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5530799

Safety Information

MSDS https://www.hit2lead.com/sds/5530799_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart