CH4005847295 ChemBridge Corporation

(2-methoxybenzyl)(2-methoxy-1-methylethyl)amine

  • MDL number MFCD01817787
  • Cas Number : 355818-30-3
  • Molecular Weight : 209.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5564802

Safety Information

MSDS https://www.hit2lead.com/sds/5564802_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart