CH4005848109 ChemBridge Corporation

3-[(2-methoxy-2-oxoethyl)amino]benzoic acid

  • MDL number MFCD00779526
  • Cas Number : 418788-94-0
  • Molecular Weight : 209.1986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5568834

Safety Information

MSDS https://www.hit2lead.com/sds/5568834_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart