CH4005848607 ChemBridge Corporation

3-fluoro-N-(4-iodo-2-methylphenyl)benzamide

  • MDL number MFCD00783364
  • Cas Number : 314022-37-2
  • Molecular Weight : 355.1461

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5571464

Safety Information

MSDS https://www.hit2lead.com/sds/5571464_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart