CH4005848889 ChemBridge Corporation

5-bromo-N-(4-chloro-2,5-dimethoxyphenyl)-2-furamide

  • MDL number MFCD00783393
  • Cas Number : 314022-39-4
  • Molecular Weight : 360.5877

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5572992

Safety Information

MSDS https://www.hit2lead.com/sds/5572992_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart