CH4005852587 ChemBridge Corporation

4-(1,3-dihydro-2H-isoindol-2-yl)benzoic acid

  • MDL number MFCD00755022
  • Cas Number : 329715-35-7
  • Molecular Weight : 239.2692

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5616316

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5616316
Availability details for
Quantity
  • Availability
  • Add to Cart