CH4005853991 ChemBridge Corporation

2-phenyl-2,5,6,7-tetrahydro-4H-1,2,3-benzotriazol-4-one

  • MDL number MFCD00168390
  • Cas Number : 97507-52-3
  • Molecular Weight : 213.2352

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5626281

Safety Information

MSDS https://www.hit2lead.com/sds/5626281_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart