CH4005856686 ChemBridge Corporation

methyl 4-(3-hydroxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00718126
  • Cas Number : 301319-88-0
  • Molecular Weight : 276.2878

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5647804

Safety Information

MSDS https://www.hit2lead.com/sds/5647804_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart