CH4005858158 ChemBridge Corporation

5-(4-chlorobenzyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00090499
  • Cas Number : 39181-43-6
  • Molecular Weight : 225.6979

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5654564

Safety Information

MSDS https://www.hit2lead.com/sds/5654564_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart