CH4005859422 ChemBridge Corporation

3-(4-iodophenyl)-6-methoxy-2H-chromen-2-one

  • MDL number MFCD01179328
  • Cas Number : 419540-74-2
  • Molecular Weight : 378.1612

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5661978

Safety Information

MSDS https://www.hit2lead.com/sds/5661978_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart