CH4005860835 ChemBridge Corporation

(5E)-5-(5-bromo-2-hydroxy-3-methoxybenzylidene)-3-ethyl-2-thioxo-1,3-thiazolidin-4-one

  • MDL number MFCD01152484
  • Cas Number : 359899-47-1
  • Molecular Weight : 374.2733

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5669084

Safety Information

MSDS https://www.hit2lead.com/sds/5669084_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart