CH4005862402 ChemBridge Corporation

1-[(1-phenyl-1H-tetrazol-5-yl)thio]acetone

  • MDL number MFCD00453350
  • Cas Number : 25803-68-3
  • Molecular Weight : 234.2776

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5676310

Safety Information

MSDS https://www.hit2lead.com/sds/5676310_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart