CH4005864526 ChemBridge Corporation

4-(4-hydroxy-6-methoxy-2-methylquinolin-3-yl)butan-2-one

  • MDL number MFCD01143178
  • Cas Number : 332150-27-3
  • Molecular Weight : 259.3004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5688348

Safety Information

MSDS https://www.hit2lead.com/sds/5688348_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart