CH4005864719 ChemBridge Corporation

4-(6-bromo-4-chloro-2-methylquinolin-3-yl)butan-2-one

  • MDL number MFCD01143185
  • Cas Number : 27311-88-2
  • Molecular Weight : 326.6161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5689881

Safety Information

MSDS https://www.hit2lead.com/sds/5689881_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart