CH4005871675 ChemBridge Corporation

(2-acetyl-4,5-dimethoxyphenyl)acetic acid

  • MDL number MFCD00230664
  • Cas Number : 38210-84-3
  • Molecular Weight : 238.2366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5733151

Safety Information

MSDS https://www.hit2lead.com/sds/5733151_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart