CH4005872033 ChemBridge Corporation

(5Z)-5-[(5-bromo-2-furyl)methylene]-3-methyl-2-thioxo-1,3-thiazolidin-4-one

  • MDL number MFCD01361727
  • Cas Number : 292024-92-1
  • Molecular Weight : 304.1834

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5735570

Safety Information

MSDS https://www.hit2lead.com/sds/5735570_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart