CH4005874705 ChemBridge Corporation

3-[(2,2-dimethylpropanoyl)amino]-2-methylbenzoic acid

  • MDL number MFCD01158917
  • Cas Number : 312317-36-5
  • Molecular Weight : 235.279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5755472

Safety Information

MSDS https://www.hit2lead.com/sds/5755472_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart