CH4005875838 ChemBridge Corporation

4-(2-nitro-5-piperazin-1-ylphenyl)morpholine

  • MDL number MFCD01173869
  • Cas Number : 332023-13-9
  • Molecular Weight : 292.3336

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5764926

Safety Information

MSDS https://www.hit2lead.com/sds/5764926_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart