CH4005876233 ChemBridge Corporation

5-(2-methylphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00652823
  • Cas Number : 59565-54-7
  • Molecular Weight : 191.2529

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5767930

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5767930
Availability details for
Quantity
  • Availability
  • Add to Cart