CH4005878801 ChemBridge Corporation

2-(4-bromophenyl)-5,7-dimethylpyrazolo[1,5-a]pyrimidine

  • MDL number MFCD00634583
  • Cas Number : 310422-24-3
  • Molecular Weight : 302.1692

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5789332

Safety Information

MSDS https://www.hit2lead.com/sds/5789332_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart