CH4005879584 ChemBridge Corporation

methyl 3-(3-bromophenyl)isoxazole-4-carboxylate

  • MDL number MFCD01655069
  • Cas Number : 267651-85-4
  • Molecular Weight : 282.0901

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5792788

Safety Information

MSDS https://www.hit2lead.com/sds/5792788_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart