CH4005880610 ChemBridge Corporation

2-amino-5-benzyl-4-methyl-3-thiophenecarboxamide

  • MDL number MFCD01445240
  • Cas Number : 57243-81-9
  • Molecular Weight : 246.3281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5800379

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5800379
Availability details for
Quantity
  • Availability
  • Add to Cart