CH4005881585 ChemBridge Corporation

3-(3-hydroxyphenyl)-3,4-dihydrobenzo[f]quinolin-1(2H)-one

  • MDL number MFCD01540875
  • Cas Number : 333759-52-7
  • Molecular Weight : 289.3279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5806011

Safety Information

MSDS https://www.hit2lead.com/sds/5806011_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart