CH4005883073 ChemBridge Corporation

2-(3-phenyl-5-isoxazolyl)phenol

  • MDL number MFCD01820980
  • Cas Number : 67139-38-2
  • Molecular Weight : 237.2533

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5810970

Safety Information

MSDS https://www.hit2lead.com/sds/5810970_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart